Skip to Content
Merck
All Photos(4)

Key Documents

HPA019220

Sigma-Aldrich

Anti-SPATA4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Spermatogenesis-associated protein 4, Anti-Testis and spermatogenesis cell-related protein 2, Anti-Testis spermatocyte apoptosis-related gene 2 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Check Cart for Availability


Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

LILLHMLQRKLGRKLNPEWFDVKPTVGEVTLNHLPAQASGRRYNLKVKRGRVVPVLPNIGSGGSSHREIHVKQAGQHSYYSAMKPIRN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPATA4(132851)

General description

SPATA4 (spermatogenesis-associated protein 4) localizes in the cytoplasm and the nucleus. It contains a DUF1042 domain. SPATA4 is expressed primarily in testis, mainly in testis interstitial, leydig cell, seminiferous tubule and germ cell. SPATA4 is also referred as TSARG2 (testis spermatocyte apoptosis-related gene 2).

Immunogen

Spermatogenesis-associated protein 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SPATA4 (spermatogenesis-associated protein 4) is an interacting partner of the TRAPP vesicle-tethering complex which is involved in protein transport. Ectopic expression of SPATA4 in MCF7 (human breast adenocarcinoma) cells allows fast progression of cells through S-pahse.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74208

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Débora Teixeira Duarte et al.
FEBS letters, 585(17), 2676-2681 (2011-08-11)
The TRAPP vesicle-tethering complex consists of more than 10 distinct polypeptides and is involved in protein transport. Using the C2 subunit as bait we identified SPATA4, a spermatocyte-specific protein of unknown function, as an interacting partner in a yeast two
Shang-feng Liu et al.
Journal of biochemistry and molecular biology, 38(6), 739-747 (2005-12-13)
Full-length cDNA sequences of four novel SPATA4 genes in chimpanzee, cow, chicken and ascidian were identified by bioinformatic analysis using mouse or human SPATA4 cDNA fragment as electronic probe. All these genes have 6 exons and have similar protein molecular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service