Skip to Content
Merck
All Photos(3)

Key Documents

HPA015716

Sigma-Aldrich

Anti-PDZD8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PDZ domain-containing protein 8, Anti-Sarcoma antigen NY-SAR-84/NY-SAR- 104

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


Estimated to ship on13 April 2025



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


Estimated to ship on13 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SCLFDIEACHRYLNIALWCRDPFKLGGLICLGHVSLKLEDVALGCLATSNTEYLSKLRLEAPSPKAIVTRTALRNLSMQKGFNDKFCYGDITIHFKYLKEGESDHHVVTNVEKEKEPHLVEEVSVLPKEEQFVGQMGLTENKHSFQDTQF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDZD8(118987)

Immunogen

PDZ domain-containing protein 8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PDZ domain containing 8 (PDZD8) is a novel moesin-interacting protein that helps in regulating the uncoating of the HIV-1 capsid. Overexpression of this gene promotes initiation of reverse transcription and infection by pseudotyped retroviruses increases whereas knockdown of this gene decreases HIV-1 infection. It serves as a positive mediator of retroviral infection and is capable of binding to the capsid proteins of different retroviruses.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72163

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Alexander Guth et al.
Journal of virology, 88(9), 4612-4623 (2014-02-21)
Following human immunodeficiency virus type 1 (HIV-1) entry into the host cell, the viral capsid gradually disassembles in a process called uncoating. A proper rate of uncoating is important for reverse transcription of the HIV-1 genome. Host restriction factors such
Keiko Morita et al.
iScience, 25(12), 105612-105612 (2022-12-06)
Dyslipidemia including the accumulation of cholesteryl esters (CEs) in the brain is associated with neurological disorders, although the underlying mechanism has been unclear. PDZD8, a Rab7 effector protein, transfers lipids between endoplasmic reticulum (ER) and Rab7-positive organelles and thereby promotes
Matthew S Henning et al.
Journal of virology, 84(17), 8990-8995 (2010-06-25)
In a yeast two-hybrid screen for cellular factors that could interact with human immunodeficiency virus type 1 (HIV-1) Gag protein, we identified PDZD8 and confirmed the interaction by coimmunoprecipitation (co-IP). PDZD8 overexpression promoted the initiation of reverse transcription and increased
Philipp Walch et al.
Cell host & microbe, 29(8), 1316-1332 (2021-07-09)
Intracellular bacterial pathogens inject effector proteins to hijack host cellular processes and promote their survival and proliferation. To systematically map effector-host protein-protein interactions (PPIs) during infection, we generated a library of 32 Salmonella enterica serovar Typhimurium (STm) strains expressing chromosomally
Shijian Zhang et al.
Virology, 481, 73-78 (2015-03-17)
PDZD8 can bind the capsid proteins of different retroviruses, and transient knockdown of PDZD8 results in a decrease in the efficiency of an early, post-entry event in the retrovirus life cycle. Here we used the CRISPR-CAS9 system to create cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service