Skip to Content
Merck
All Photos(3)

Key Documents

HPA015275

Sigma-Aldrich

Anti-SLC30A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Solute carrier family 30 member 1, Anti-Zinc transporter 1, Anti-ZnT-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$1,040.00

$1,040.00


In StockDetails



Select a Size

Change View
100 μL
$1,040.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$1,040.00


In StockDetails


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

HHSGFSQDSGHGHSHGGHGHGHGLPKGPRVKSTRPGSSDINVAPGEQGPDQEETNTLVANTSNSNGLKLDPADPENPRSGDTVEVQVNGNLVREPDHMELEEDRAGQLNMRGV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC30A1(7779)

General description

SLC30A1 (solute carrier family 30, member 1) is a transmembrane transporter of the zinc-regulating family of proteins. It has a wide range of tissue expression, with predominant expression in brain, heart, pancreas and testis. It is the first ZnT (zinc transporter) family member to be identified, and the ZnT family in turn is a part of cation diffusion facilitator (CDF) family of proteins. This protein contains six transmembrane domains which are connected by histidine rich loops.

Immunogen

Zinc transporter 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SLC30A1 (solute carrier family 30, member 1) is involved in the metabolism of zinc, and prevents cellular zinc toxicity. It is a newly recognized endogenous LTCC (L-type calcium channel)-inhibitor, where it interacts with the LTCCβ subunit. This prevents the transport of LTCCα1 subunit to the plasma membrane. In heart, the inhibition of LTCC affects the cardiac electrical remodeling post-atrial fibrillation. In brain, it influences the synaptic responses. It increases the surface expression and activity of T-type calcium channels such as, CaV3.1 and CaV3.2, by activating the Ras-ERK signaling pathway. Inactivation of this gene leads to death in early embryonic stage. It controls Ras-mediated signaling, by interacting with and activating Raf-1. The expression of this protein is abnormal in the brains of patients with mild cognitive impairment (MCI), and early and late stage Alzheimer′s disease (AD). Thus, pathogenesis of AD might be associated with aberrations in zinc homeostasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73291

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shiri Levy et al.
The Journal of biological chemistry, 284(47), 32434-32443 (2009-09-22)
The L-type calcium channel (LTCC) has a variety of physiological roles that are critical for the proper function of many cell types and organs. Recently, a member of the zinc-regulating family of proteins, ZnT-1, was recognized as an endogenous inhibitor
Eden Shusterman et al.
Metallomics : integrated biometal science, 6(9), 1656-1663 (2014-06-22)
ZnT-1 is a Cation Diffusion Facilitator (CDF) family protein, and is present throughout the phylogenetic tree from bacteria to humans. Since its original cloning in 1995, ZnT-1 has been considered to be the major Zn(2+) extruding transporter, based on its
Merav Mor et al.
American journal of physiology. Cell physiology, 303(2), C192-C203 (2012-05-11)
Zinc transporter-1 (ZnT-1) is a putative zinc transporter that confers cellular resistance from zinc toxicity. In addition, ZnT-1 has important regulatory functions, including inhibition of L-type calcium channels and activation of Raf-1 kinase. Here we studied the effects of ZnT-1
Yoram Etzion et al.
Journal of cardiovascular electrophysiology, 19(2), 157-164 (2007-11-01)
Until recently, the membrane protein ZnT-1 was studied mainly in the context of zinc homeostasis. However, new findings indicate that it acts as an inhibitor of L-type calcium channels. We recently found that acute rapid pacing of the rat atria
Ganna Lyubartseva et al.
Brain pathology (Zurich, Switzerland), 20(2), 343-350 (2009-04-18)
Our previous studies demonstrate alterations of zinc (Zn) transporter proteins ZnT-1, ZnT-4 and ZnT-6 in vulnerable brain regions of subjects with mild cognitive impairment (MCI), and early and late stage Alzheimer's disease (AD), suggesting disruptions of Zn homeostasis may play

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service