Skip to Content
Merck
All Photos(2)

Key Documents

MABT522

Sigma-Aldrich

Anti-TUBA1B (alpha-tubulin ubiquitous) Antibody

rabbit monoclonal, RM113

Synonym(s):

Tubulin alpha-1B chain, Alpha-tubulin ubiquitous, Tubulin K-alpha-1, Tubulin alpha-ubiquitous chain, alpha-Tubulin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$904.00

$904.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$904.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$904.00


Estimated to ship on02 June 2025


Request a Bulk Order

Product Name

Anti-alpha-Tubulin Antibody, clone RM113, clone RM113, from rabbit

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM113, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

General description

Microtubules are involved in a wide variety of cellular activities ranging from mitosis and transport events to cell movement and the maintenance of cell shape. Tubulin itself is a globular protein which consists of two polypeptides (alpha and beta tubulin). Alpha and beta tubulin dimers are assembled to 13 protofilaments that form a microtubule of 22 nm diameter. Tyrosine ligase adds a C-terminal tyrosin to monomeric alpha tubulin. Assembled microtubules can be detyrosinated by a cytoskeleton associated carboxypeptidase. Detyrosinated alpha tubulin is referred to as Glu-tubulin. Another post-translational modification of detyrosinated alpha tubulin is C-terminal polyglutamylation which is characteristic for microtubules in neuronal cells and the mitotic spindle.

Immunogen

A peptide corresponding to the C-terminus of α-Tubulin.

Application

Immunocytochemistry Analysis (ICC): A 1:200 dilution from a representative lot detected alpha-Tubulin in A431 cells.

This Anti-alpha-Tubulin Antibody, clone RM113 is validated for use in Western Blotting, Immunocytochemistry for the detection of alpha-Tubulin.

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected alpha-Tubulin in A431 cell lysate.

Target description

~53 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Yu et al.
Frontiers in cell and developmental biology, 10, 918235-918235 (2023-01-03)
SATB homeobox proteins are important regulators of developmental gene expression. Among the stem cell lineages that emerge during early embryonic development, trophoblast stem (TS) cells exhibit robust SATB expression. Both SATB1 and SATB2 act to maintain the trophoblast stem-state. However

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service