Skip to Content
Merck
All Photos(7)

Key Documents

HPA013742

Sigma-Aldrich

Anti-SLCO2A1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MATR1, Anti-OATP2A1, Anti-PGT, Anti-SLC21A2, Anti-solute carrier organic anion transporter family, member 2A1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$952.00

$952.00


Check Cart for Availability


Select a Size

Change View
100 μL
$952.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$952.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

PSTSSSIHPQSPACRRDCSCPDSIFHPVCGDNGIEYLSPCHAGCSNINMSSATSKQLIYLNCSCVTGGSASAKTGSCPVPCAH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLCO2A1(6578)

General description

The gene SLCO2A1 (solute carrier organic anion transporter family, member 2A1) is mapped to human chromosome 3q21 and contains 14 exons that encode a 643 amino acid protein. This organic anion cell-surface transporter is predominantly expressed in several peripheral tissues and in the brain.

Immunogen

solute carrier organic anion transporter family, member 2A1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene SLCO2A1 (solute carrier organic anion transporter family, member 2A1) encodes a prostaglandin (PG) transporter that belongs to the family of 12-membrane-spanning transporters. It may function in the efflux of newly synthesized PGs from cells, epithelial PG transport, and in the clearance and degradation of PG.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72631

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoko Yamaguchi et al.
Intestinal research, 16(3), 393-399 (2018-08-10)
We recently identified recessive mutations in the solute carrier organic anion transporter family member 2A1 gene (SLCO2A1) as causative variants of chronic enteropathy associated with SLCO2A1 (CEAS). The aim of this study was to evaluate SLCO2A1 protein expression in the
Junji Umeno et al.
PLoS genetics, 11(11), e1005581-e1005581 (2015-11-06)
Previously, we proposed a rare autosomal recessive inherited enteropathy characterized by persistent blood and protein loss from the small intestine as chronic nonspecific multiple ulcers of the small intestine (CNSU). By whole-exome sequencing in five Japanese patients with CNSU and
Yimin Dai et al.
Orphanet journal of rare diseases, 19(1), 229-229 (2024-06-12)
Chronic enteropathy associated with SLCO2A1 gene (CEAS) results from loss-of-function variants in SLCO2A1, which encodes the prostaglandin transporter (PGT). CEAS follows an autosomal recessive inheritance pattern. To date, approximate 30 pathogenic variants have been reported in CEAS. We performed whole
Chizuru Ariake et al.
Internal medicine (Tokyo, Japan), 61(17), 2607-2611 (2022-02-22)
Chronic enteropathy associated with solute carrier organic anion transporter family, member 2A1 (SLCO2A1) (CEAS) is a rare autosomal recessive hereditary disease characterized by chronic persistent anemia and hypoproteinemia. Its diagnosis typically requires a genetic analysis. The efficacy of immunohistochemical staining
Keisuke Jimbo et al.
PloS one, 15(11), e0241869-e0241869 (2020-11-10)
Chronic enteropathy associated with SLCO2A1 gene (CEAS) is caused by loss-of-function mutations in SLCO2A1, which encodes a prostaglandin (PG) transporter. In this study, we report a sibling case of CEAS with a novel pathogenic variant of the SLCO2A1 gene. Compound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service