Skip to Content
Merck
All Photos(2)

Key Documents

MTOX1302

Sigma-Aldrich

Canine MDR1 Knockout, Human BCRP/OATP2B1 double Knockin MDCKII Cells

canine kidney (cocker spaniel)

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
$2,450.00

$2,450.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
$2,450.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

$2,450.00


Check Cart for Availability

Request a Bulk Order

Product Name

Canine MDR1 Knockout, Human BCRP/OATP2B1 double Knockin MDCKII Cells,

biological source

canine kidney (cocker spaniel)

form

liquid

shipped in

dry ice

storage temp.

−196°C

Gene Information

human ... ABCG2(9429)

General description

MDCK II - Madin-Darby canine kidney- is a subclone derived from the heterogenous The canine MDR1 (cP-gp) efflux transporter gene has been effectively disrupted in both alleles. There is no expression of the cP-gp. Human BCRP and Human OATP2B1 has been transfected into the MDCKII cell line. Validation studies have shown BCRP dependent efflux of estrone-3 sulfate (requires OATP2B1) and teriflunomide (does not require OATP2B1).

Features and Benefits

MDCK II - Madin-Darby canine kidney- is a subclone derived from the heterogenous parent line - MDCK (ECACC catalogue no. 85011435) MDCK II cells, which predominate in later passages from MDCK, are reported to display electrical resistance of 100 ohm/cm2. This strain is thought to be derived from the distal tubule or collecting duct of the nephron. The cell line can be used as an experimental model to study the generation and maintenance of cell surface polarity in epithelial cells.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service