Skip to Content
Merck
All Photos(4)

Key Documents

HPA007120

Sigma-Aldrich

Anti-STK3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AP003355.2, Anti-Serine/threonine-protein kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

ATSTMSEGAQTMIEHNSTMLESDLGTMVINSEDEEEEDGTMKRNATSPQVQRPSFMDYFDKQDFKNKSHENCNQNMHEPFPMSKNVFPDNWKVPQDGDFDFLKNLSLEELQMRLKALDPMMEREI

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STK3(6788)

Immunogen

serine/threonine-protein kinase 3 (EC 2.7.11.1)(STE20-like kinase MST2)(MST-2)(Mammalian STE20-like protein kinase 2)(Serine/threonine-protein kinase Krs-1) recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

STK3 (serine/threonine kinase 3) encodes a serine/threonine protein kinase. It is highly expressed in the adult kidney, skeletal and placental tissues and at very low levels in adult heart, lung, liver and brain tissues. It is activated by trans-autophosphorylation at its activation loop. It is associated with the stress-dependent cardiac hypertrophy and remodeling in the human heart. It also suppresses the Wnt/β-catenin pathway through the Hippo pathway. In association with other cytoplasmic protein kinase, STK3 regulates cellular activities during quiescence and quiescence-related apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70211

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Xu et al.
The Biochemical journal, 458(1), 159-169 (2013-11-05)
The Hippo signalling pathway can suppress the Wnt/β-catenin signalling pathway through the last downstream effectors YAP (Yes-associated protein)/TAZ (tafazzin). MST (mammalian sterile 20-like kinase) 1 functions as the upstream kinase of the Hippo pathway, and CK1ε (casein kinase 1ε) plays
Min Zi et al.
The Journal of biological chemistry, 289(35), 24275-24288 (2014-07-19)
The Hippo signaling pathway has recently moved to center stage in cardiac research because of its key role in cardiomyocyte proliferation and regeneration of the embryonic and newborn heart. However, its role in the adult heart is incompletely understood. We
H C Wang et al.
The Journal of biological chemistry, 275(33), 25850-25857 (2000-06-07)
We have identified a cell quiescence-specific 33-kDa cytoplasmic protein kinase (p33(QIK), Quiescence-Induced Kinase) based on induction of p33(QIK)-specific kinase activity of cells growth-arrested in the quiescent phase and deactivation upon entry into the cell cycle. Blockage of macromolecular synthesis prevents
C L Creasy et al.
Gene, 167(1-2), 303-306 (1995-12-29)
We have identified a second human homology of the yeast Ste20 protein kinase family, which we designate MST2. MST2 is most similar to the previously identified MST1 protein kinase (78% identity, 88% similarity). Northern analysis indicates that MST2 mRNA is
Lisheng Ni et al.
Structure (London, England : 1993), 21(10), 1757-1768 (2013-08-27)
The tumor-suppressive Hippo pathway controls tissue homeostasis through balancing cell proliferation and apoptosis. Activation of the kinases Mst1 and Mst2 (Mst1/2) is a key upstream event in this pathway and remains poorly understood. Mst1/2 and their critical regulators RASSFs contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service