Skip to Content
Merck
All Photos(6)

Key Documents

HPA004902

Sigma-Aldrich

Anti-CBX3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Chromobox protein homolog 3, Anti-HECH, Anti-HP1 gamma, Anti-Heterochromatin protein 1 homolog gamma, Anti-Modifier 2 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RVVNGKVEYFLKWKGFTDADNTWEPEENLDCPELIEAFLNSQKAGKEKDGTKRKSLSDSESDDSKSKKKRDAADKPRGFARGLDPERIIGATDSSGELMFLMKWKDSDEA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CBX3(11335)

General description

CBX3 (chromobox homolog 3), a heterchromatin protein, is a member of heterochromatin protein 1 family (HP1).

Immunogen

Chromobox protein homolog 3 recombinant protein epitope signature tag (PrEST)

Application

Anti-CBX3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CBX3 (chromobox homolog 3) is majorly responsible for transcriptional regulation (co-transcriptional RNA processing), formation and packaging of heterochromatin. It also participates in DNA repair, gene silencing (RNAi-mediated silencing) and telomeric function. Studies have suggested that CBX3 acts as a prognostic biomarker in cancer development. An elevated expression has been reported in the non-small cell lung cancer (NSCLC). It has also been reported that CBX3 plays a vital role in the chromatin-mediated repression of the HIV-1 gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86943

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Yan et al.
PloS one, 8(2), e55790-e55790 (2013-02-15)
Cancer cell metabolism responsive to androgen deprivation therapy (ADT) may be involved in the development and progression of prostate cancer and the ultimate failure of androgen-deprivation therapy. To investigate the metabolism regulation effects on androgen-independent growth of prostate cancer, an
Ji Zhou et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 9777-9785 (2014-07-02)
Heterochromatin protein 1γ (HP1γ), which binds to di- or trimethylated lysine 9 on histone H3 (H3K9), plays an important role in chromatin packaging and gene transcriptional regulation. Recently, HP1γ has been implicated in cancer development. However, its clinical relevance and
Andrea Smallwood et al.
Genome research, 22(8), 1426-1436 (2012-06-12)
CBX5, CBX1, and CBX3 (HP1α, β, and γ, respectively) play an evolutionarily conserved role in the formation and maintenance of heterochromatin. In addition, CBX5, CBX1, and CBX3 may also participate in transcriptional regulation of genes. Recently, CBX3 binding to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service