Skip to Content
Merck
All Photos(3)

Key Documents

MABT217

Sigma-Aldrich

Anti-Calreticulin Antibody, clone 16B11.1

clone 16B11.1, from mouse

Synonym(s):

Calreticulin, CRP55, Calregulin, Endoplasmic reticulum resident protein 60, ERp60, HACBP, grp60

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$778.00

$778.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1681

Request a Bulk Order

Select a Size

Change View
100 μL
$778.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$778.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1681

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

16B11.1, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CALR(811)

Related Categories

General description

Calreticulin is a molecular Ca+ binding chaperone promoting folding, oligomeric assembly, and quality control in ER. Calreticulin is a highly versatile lectin-like chaperone, and it participates during the synthesis of a variety of molecules, including ion channels, surface receptors, integrins and transporters. Calreticulin indeed provides up to 45% of the Ca2+-buffering capacity for a pool of the IP3-sensitive Ca2+ inside the ER. Calreticulin also functions in cell adhesion, integrin-dependent Ca2+ signaling and steroid-sensitive gene expression.

Immunogen

GST-tagged recombinant protein corresponding to human Calreticulin.

Application

Immunohistochemistry Analysis: A 1:300-1,000 dilution from a representative lot detected Calreticulin in human colorectal cancer and human tonsil tissues.

Flow Cyometry Analysis: A 1:500 dilution from a representative lot detected Calreticulin in non serum starved HeLa cells.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-Calreticulin Antibody, clone 16B11.1 is validated for use in Western Blotting, IHC, Flow Cytometry for the detection of Calreticulin.

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Calreticulin on 10 µg of HeLa cell lysate.

Target description

~48 kDa observed

Linkage

Replaces: MABT145

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service