Skip to Content
Merck
All Photos(3)

Key Documents

HISTO-AB

Sigma-Aldrich

Visikol® HISTO Antibody Buffer

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$138.00
1 EACH
$251.00

$138.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$138.00
1 EACH
$251.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.47

$138.00


Check Cart for Availability

shipped in

ambient

storage temp.

2-8°C

General description

Visikol HISTO Antibody Buffer is an antigen retrieval buffer designed to aid in visualization of tissues in 3D. Using the Visikol HISTO Antibody Buffer gives researchers more time to focus on what matters, instead of preparing media.

Application

Ready-to-use antigen retrieval agent in Visikol HISTO tissue clearing protocols.

Features and Benefits

  • Render tissue samples transparent for imaging.
  • Achieve clearer, sharper images in antibody tissue staining applications.
  • The Visikol HISTO tissue clearing process does not affect prior immunolabeling.
  • Save time. The Visikol HISTO method is rapid relative to many popular current tissue clearing techniques.
  • Retain morphology and lipid content in your samples.
  • The HISTO tissue clearing method is easily reversible for follow up 2D histology/IHC or for 3D data validation against traditional histology.
  • Non-toxic.

Legal Information

HISTO is a trademark of Visikol, Inc.
Visikol is a registered trademark of Visikol, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Visikol® HISTOTM tissue clearing solutions enable developmental biology, neuroscience, and cancer research with reversible protocols.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service