Key Documents
E4140
Sigma-Aldrich
Embedding molds
19 Flat bullet-shaped unindexed cavities, clear
Select a Size
1 EA
$46.40
$46.40
Check Cart for Availability
Select a Size
Change View
1 EA
$46.40
About This Item
UNSPSC Code:
41116155
NACRES:
MA.01
$46.40
Check Cart for Availability
material
silicone
color
clear
General description
With cavities for the preparation of microscopy samples. Two variations are available. Maroon molds contain additives for chemical and thermal resistance; they are durable and economical. Clear molds are used where optical clarity is necessary for precise sample alignment; recommended for use with low-temperature UV-cured resins.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. For Product E4390 What is a depth of cavities is it 4 mm?
    1 answer
    1. The bullet shape cavities are 7 mm wide x 14 mm long x 4 mm deep.
      Helpful?