Skip to Content
Merck
All Photos(2)

Documents

H7625

Sigma-Aldrich

D-Histidine monohydrochloride monohydrate

≥98% (TLC)

Synonym(s):

D-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4161356
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

product name

D-Histidine monohydrochloride monohydrate, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

color

white

mp

254 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

O.Cl.N[C@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m1../s1

InChI key

CMXXUDSWGMGYLZ-ZJIMSODOSA-N

Gene Information

human ... HRH1(3269)
mouse ... HRH1(15465)
rat ... HRH1(24448)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Unnatural isomer of histidine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Champe PC
Biochemistry (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service