Skip to Content
Merck
All Photos(1)

Key Documents

H0755000

Histidine hydrochloride monohydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Histidine monohydrochloride monohydrate, L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$328.00

$328.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 MG
$328.00

About This Item

Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Beilstein:
4168261
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$328.00


Available to ship on15 April 2025Details


Request a Bulk Order

grade

pharmaceutical primary standard

API family

histidine

manufacturer/tradename

EDQM

mp

254 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.Cl.N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1

InChI key

CMXXUDSWGMGYLZ-XRIGFGBMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Histidine hydrochloride monohydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Saller et al.
Journal of pharmaceutical sciences, 104(4), 1440-1450 (2015-01-22)
In a typical manufacturing setup for biopharmaceutical drug products, the fill and dosing pump is placed after the final sterile filtration unit in order to ensure adequate dispensing accuracy and avoid backpressure peaks. Given the sensitivity of protein molecules, peristaltic
Aldo Roda et al.
The Analyst, 139(24), 6494-6501 (2014-10-25)
Increasingly, smartphones are used as portable personal computers, revolutionizing communication styles and entire lifestyles. Using 3D-printing technology we have made a disposable minicartridge that can be easily prototyped to turn any kind of smartphone or tablet into a portable luminometer
Nam Ah Kim et al.
Drug development and industrial pharmacy, 41(2), 300-306 (2014-02-08)
Correlation of thermodynamic and secondary structural stability of proteins at various buffer pHs was investigated using differential scanning calorimetry (DSC), dynamic light scattering (DLS) and attenuated total reflection Fourier-transform infrared spectroscopy (ATR FT-IR). Recombinant human epithelial growth factor (rhEGF) was
Tino Jaschinski et al.
Rapid communications in mass spectrometry : RCM, 28(13), 1521-1529 (2014-05-28)
Laser desorption ionization for mass spectrometric measurements (LDI MS) is supported by nanostructured materials. This technique helps to overcome known limitations of matrix-assisted laser desorption/ionization (MALDI) and especially avoids interfering signals caused by matrix components. LDI can be supported by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service