Skip to Content
Merck
All Photos(1)

Key Documents

H6411

Sigma-Aldrich

Monoclonal Anti-HIF-1α antibody produced in mouse

~1 mg/mL, clone OZ12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Hypoxia Inducible Factor 1, α subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$568.00

$568.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$568.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$568.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

OZ12, monoclonal

form

buffered aqueous solution

mol wt

antigen 120 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 2 μg using 1 mg protein lysate
indirect immunofluorescence: 2 μg/mL

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIF1A(3091)

Immunogen

peptide corresponding to amino acids 530-826 of the human HIF-1α protein.

Physical form

Solution in phosphate buffered saline pH 7.4, containing 0.08% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Arnesen et al.
The Biochemical journal, 386(Pt 3), 433-443 (2004-10-22)
Protein acetyltransferases and deacetylases have been implicated in oncogenesis, apoptosis and cell cycle regulation. Most of the protein acetyltransferases described acetylate epsilon-amino groups of lysine residues within proteins. Mouse ARD1 (homologue of yeast Ard1p, where Ard1p stands for arrest defective

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service