GS61
GST A2-2, Recombinant Human
Synonym(s):
GST2, GTA2, GTH2, glutathione S-transferase alpha 2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
Assay
>95% (SDS-PAGE)
form
frozen liquid
specific activity
15.95 units/mg protein
mol wt
23 kDa
concentration
1.52 mg/mL
storage temp.
−70°C
Gene Information
human ... GSTA2(2939)
General description
using spectrophotometric determination of 1-chloro-2,4-dinitrobenzene (CDNB) conjugation with reduced glutathione (1 mM) in 100 mM NaPO4 (pH 6.5) at 30°C.
Biochem/physiol Actions
Glutathione S-transferase alpha 2 (GSTA2) is an enzyme that in humans is encoded by the GSTA2 gene. Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. The GSTs are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.
The genes encoding these enzymes are known to be highly polymorphic. These genetic variations can change an individual′s susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of some drugs. This gene encodes a glutathione S-tranferase belonging to the alpha class. The alpha class genes, located in a cluster mapped to chromosome 6, are the most abundantly expressed glutathione S-transferases in liver. In addition to metabolizing bilirubin and certain anti-cancer drugs in the liver, the alpha class of these enzymes exhibit glutathione peroxidase activity thereby protecting the cells from reactive oxygen species and the products of peroxidation.
Storage and Stability
The enzyme should be used by the end-user customer within 1 year of receipt.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Haematologica, 99(1), 172-179 (2013-09-24)
Busulfan liver metabolism depends on glutathione, a crucial mediator of cellular and systemic stress. Here we investigated 40 polymorphisms at 27 loci involved in hepatic glutathione homeostasis, with the aim of testing their impact on the clinical outcome of 185
Drug metabolism and pharmacokinetics, 26(6), 646-658 (2011-08-17)
Glutathione S-transferases (GSTs) play a vital role in the phase II biotransformation of many chemicals, including anticancer drugs. In this study, to elucidate the haplotype structures of the two closely related alpha-class genes GSTA1 and GSTA2, we screened for genetic
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(8), 3198-3208 (2013-05-08)
Steroidogenic factor 1 (SF-1) is a master regulator for steroidogenesis. In this study, we identified novel SF-1 target genes using a genome-wide promoter tiling array and a DNA microarray. SF-1 was found to regulate human glutathione S-transferase A (GSTA) family
Oncology reports, 22(3), 593-598 (2009-07-30)
Glutathione-S-transferases (GSTs) are a super-family of phase II metabolizing enzymes that catalyse the detoxification of a large range of endogenous and exogenous toxic compounds, playing an important role in protecting cells against damage, through glutathione conjugation with electrophilic substances. Polymorphic
Journal of cellular biochemistry, 95(2), 339-351 (2005-03-22)
The effectiveness of bifunctional alkylating nitrogen mustard compounds in chemotherapy is related to their ability to form DNA inter-strand crosslinks. Patients exposed to DNA inter-strand crosslinking (ICL) agents subsequently experience an elevated incidence of myelodysplastic syndromes (MDS) and MDS related
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service