Skip to Content
Merck
All Photos(1)

Key Documents

GERPN1051-2ML

Streptavidin-Biotinylated HRP Complex

Cytiva RPN1051-2ML

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$685.00

$685.00


Check Cart for Availability


Select a Size

Change View
2 ML
$685.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$685.00


Check Cart for Availability

packaging

pkg of 2 mL

manufacturer/tradename

Cytiva RPN1051-2ML

color

Colorless

shipped in

wet ice

storage temp.

2-8°C

General description

Streptavidin-Biotinylated Horseradish Peroxidase Complex.

Application

The conjugation of horseradish peroxidase and streptavidin is achieved using the periodate coupling method described by Nakane and coworkers. This product is sold on the basis of titer in an ELISA and performance in a Western blot. As a result the final Protein concentration of the conjugate (RPN 1231) will vary only minimally between batches.

Biotin is linked via a spacer arm to horseradish peroxidase under conditions that preserve high enzyme activity and give a high degree of biotinylation as judged by its ability to bind to streptavidin. Excess labeling reagent is removed by gel filtration.

Biotinylated enzyme is added to streptavidin to form complexes. The complex is optimized to produce an excellent combination of titer and sensitivity for binding to biotinylated Protein. This product is sold on the basis of titer determined in a standard assay. As a result the final Protein concentration of the complex may vary minimally between batches.

Features and Benefits

  • Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications.
  • High specificity reagents.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service