Skip to Content
Merck
All Photos(1)

Key Documents

GE80-6501-23

Protease Inhibitor Mix

Cytiva 80-6501-23

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$498.00

$498.00


Check Cart for Availability


Select a Size

Change View
1 PKG
$498.00

About This Item

UNSPSC Code:
41105323
NACRES:
NA.32
solubility:
water: soluble
storage temp.:
−20°C

$498.00


Check Cart for Availability

feature

wetted part: no

packaging

pkg of 1 mL

manufacturer/tradename

Cytiva 80-6501-23

solubility

water: soluble

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Protease inhibitor mix.

Application

Sample preparation often requires the inhibition of protease activity. Cytiva offers this combination of competitive and noncompetitive protease inhibitors, which protect Proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.

Features and Benefits

  • Protease inhibitor cocktail specifically developed for sample preparation in 2-D studies
  • No EDTA is used, which allows optimal nuclease activity for removing nucleic acids from samples
  • Contains a mixture of both competitive and noncompetitive protease inhibitors that inhibit serine, cysteine, and calpain proteases.Optionally, EDTA may be added to inhibit metalloproteases.
  • Optimized concentration for excellent inhibition of protease activities.
  • Effectively inhibits over 95% of protease activity.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Mota-Martorell et al.
GeroScience, 42(4), 1157-1173 (2020-06-25)
Species longevity varies significantly across animal species, but the underlying molecular mechanisms remain poorly understood. Recent studies and omics approaches suggest that phenotypic traits of longevity could converge in the mammalian target of rapamycin (mTOR) signalling pathway. The present study
Xiaojing Wang et al.
Scandinavian journal of immunology, 90(3), e12791-e12791 (2019-05-28)
The epoxyeicosatrienoic acids (EETs) are products of cytochrome P450 epoxygenases and have recently been found to have an anti-inflammatory activity. However, the role of EETs in non-alcoholic steatohepatitis has not been fully understood. In this study, we investigated the protective
Natalia Mota-Martorell et al.
Redox biology, 34, 101539-101539 (2020-05-01)
Mitochondrial reactive oxygen species (ROS) production, specifically at complex I (Cx I), has been widely suggested to be one of the determinants of species longevity. The present study follows a comparative approach to analyse complex I in heart tissue from

Protocols

Protease Inhibitor Mix from Cytiva inhibits serine, cysteine, metalloproteases, and calpain proteases and is suitable for animal and plant tissues, yeast, and bacteria.

Rules and good practice in sample preparation for Western blot sample preparation from cell culture and tissue samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service