Documents
GE29-0220-94
ÄKTA® start protein purification system
AC/DC input 100 - 240 V AC (US and schuko plugs)
About This Item
UNSPSC Code:
41105300
NACRES:
NA.55
description
2 buffer inlets
manufacturer/tradename
Cytiva 29-0220-94
parameter
(single wave length)
0.5 Mpa max. pressure (operating)
0.5 Mpa max. pressure (operating, sample pump)
0.5-5 mL/min flow rate
pathlength
2 mm , flow cell
viscosity
0.6-5 cP
General description
ÄKTA start® is a preparative chromatography system for laboratory scale protein purification. ÄKTA start® is designed as a stand-alone system, with intuitive design, simple flow path, and user-friendly interface. With ÄKTA start® it is easy to purify a wide variety of proteins using built-in quick start methods or predefined templates, or by creating your own methods.
ÄKTA start® can be combined with the Frac30 fraction collector, the user-friendly UNICORN® start control software, and application-focused prepacked columns for an automated solution.
With ÄKTA start®, protein purification is quick, reliable, and easy. The system is easy to learn and use, and has an intuitive design, front-facing work area, visible flow path, and a user-friendly interface. ÄKTA start® makes it easy to purify proteins, with predefined quick start and editable method templates, real-time run control and visualization, and single-click operations with a compact and portable size suitable for cold-room operations. With proven and reliable technology like LED-based UV detection, and built-in help and diagnostics, you can trust ÄKTA start® to help you make an effortless switch from manual to automated protein purification.
Features and Benefits
  • Compact solution for quick and reliable one-step protein purification
  • Sample injection and fraction collection options
  • Quick start methods and templates for common purification techniques
  • Intuitive touchscreen display and real-time monitoring
Other Notes
This material does not include fraction collector and software.
Legal Information
Unicorn is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva
recommended