Skip to Content
Merck
All Photos(1)

Key Documents

GE10600003

Amersham Protran® Premium Western blotting membranes, nitrocellulose

pore size 0.45 μm, roll W × L 300 mm × 4 m, pkg of 1 ea

Synonym(s):

Western blotting membrane, 0.2 NC nitrocellulose membrane

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$467.00

$467.00


Check Cart for Availability


Select a Size

Change View
1 EA
$467.00

About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

$467.00


Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600003

roll W × L

300 mm × 4 m

pore size

0.45 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitrocellulose blotting membrane, provides excellent sensitivity, resolution, and low background in fluorescence Western blotting detection applications.

Application

Amersham Protran® Premium Western blotting membranes, nitrocellulose may be used in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Offers high binding capacity (162 - 180 μg IgG/cm2).
  • Suitable for proteins of different molecular weights
  • Unsupported, 100% pure nitrocellulose membrane is suitable for all ECL Western blotting systems and protein blotting applications.
  • Popular precut sizes are available for laboratory application
  • Provides excellent sensitivity, resolution and low background particularly when used with the fluorescence Amersham ECL Plex Western blotting detection system

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina Aznaourova et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(16), 9042-9053 (2020-04-04)
RNA has been proposed as an important scaffolding factor in the nucleus, aiding protein complex assembly in the dense intracellular milieu. Architectural contributions of RNA to cytosolic signaling pathways, however, remain largely unknown. Here, we devised a multidimensional gradient approach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service