GE27-2076-01
Guanosine 5′-Triphosphate, 100 mM solution (GTP)
Cytiva 27-2076-01
Select a Size
$128.00
Select a Size
About This Item
$128.00
Recommended Products
form
liquid
packaging
pkg of 25 μmol
manufacturer/tradename
Cytiva 27-2076-01
concentration
100 mM
color
Colorless
shipped in
dry ice
storage temp.
−20°C
General description
Features and Benefits
- Each lot is tested for ribonuclease contamination to ensure success in your application.
- Supplied in purified water as 100 mM solutions (pH 7.5).
- Extensively purified to yield a minimum of 99% in triphosphate form for solution (98% for lyophilized) to enhance incorporation and yield optimal experimental results.
- Available either as individual products or in a convenient set containing ATP, CTP, GTP, and UTP in separate tubes.
Storage and Stability
Analysis Note
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service