Skip to Content
Merck
All Photos(1)

Key Documents

20-177

Sigma-Aldrich

100X GDP, 100mM

Synonym(s):

100x Guanosine 5′-Diphosphate solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
$339.00

$339.00


Estimated to ship on27 May 2025



Select a Size

Change View
50 μL
$339.00

About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.41

$339.00


Estimated to ship on27 May 2025


form

liquid

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity binding assay: suitable (G-protein)

shipped in

dry ice

Application

For use as a negative loading control in Activation Assay Kits
Research Category
All

Physical form

sterile water

Storage and Stability

1 year at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seung-Min Shin et al.
Science advances, 6(3), eaay2174-eaay2174 (2020-01-31)
Oncogenic RAS mutant (RASMUT) proteins have been considered undruggable via conventional antibody regimens owing to the intracellular location restricting conventional-antibody accessibility. Here, we report a pan-RAS-targeting IgG antibody, inRas37, which directly targets the intracellularly activated form of various RASMUT subtypes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service