Skip to Content
Merck
All Photos(1)

Key Documents

GE18112766

Fill Port

ÄKTA® Design, Cytiva 18-1127-66

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500

description

Ethylenetetra fluoroethylene (ETFE), Titanium (Ti)
For use with all aqueous solutions and organic solvents commonly employed in chromatography. The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane and 100% Tetrahydrofuran.

feature

Wetted Materials: Ethylenetetra fluoroethylene (ETFE)
ÄKTA® Design

manufacturer/tradename

Cytiva 18-1127-66

parameter

10 bar max. pressure
40 °C max. temp.

diam.

(0.7 mm)

long term

1-13

Related Categories

General description

A range of manual and automated valves to meet a variety of performance requirements. Designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.

Legal Information

ÄKTA is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service