Documents
GBL204064
Grace Bio-Labs FlexWell removable incubation chambers
L × W × thickness 3.5 mm × 3.5 mm × 1.8 mm, black silicone, adhesive one side, wells 64
Synonym(s):
flexible silicone slide gasket, silicone multiwell gasket
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
black silicone
sterility
non-sterile
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 204064
L × W × thickness
3.5 mm × 3.5 mm × 1.8 mm
well area
0.12 cm2
wells
64
General description
FlexWell incubation chambers provide a convenient way to conduct binding reactions on protein microarrays using nitrocellulose coated ONCYTE® Film slides.
  • Silicone FlexWell forms wells on slides using a clean release adhesive to isolate up to 16 specimens per slide
  • Slides and FlexWell fit into a reusable slide tray organizing wells into the footprint of a SMS compliant microtiter plate
  • Gaskets may be sealed for mixing and to prevent evaporation
Legal Information
FlexWell is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.