Skip to Content
Merck
All Photos(2)

Key Documents

GBL104112

Grace Bio-Labs SecureSlip silicone supported coverglass

L × W 12 mm × 12 mm

Synonym(s):

glass coverslips, silicone backed coverglass

Sign Into View Organizational & Contract Pricing

Select a Size

75 EA
$864.00

$864.00


Check Cart for Availability


Select a Size

Change View
75 EA
$864.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

$864.00


Check Cart for Availability

material

borosilicate glass cover
colorless
silicon chamber

sterility

(sterile)
sterile

feature

wells 15 (in each sealed pack)

manufacturer/tradename

Grace Bio-Labs 104112

packaging

pkg of (Shipped as 5 sealed packs each containing 15 chambers)

L × W

12 mm × 12 mm

external L × W

130 mm × 72 mm

well area

1.44 cm2

wells

15

Looking for similar products? Visit Product Comparison Guide

General description

The SecureSlip coverglass is affixed to a thin microscopically transparent silicone base which secures to culture vessels by an electrostatic charge, not an adhesive, to prevent movement during cultivation. The silicone also acts as a hydrophobic barrier to isolate liquid reagents on glass. Individual coverglass with silicone backing can be adhered to a petri dish or plate.

These can be easily removed one at a time from the dish during culture. Once removed, coverglass can be affixed to a microscope slide by means of the tacky silicone backing for routine processing, or placed cell side down onto a drop of reagent using the silicone backing as a cover to prevent evaporation.

  • Silicone backing peels off easily for conventional mounting on microscope slides
  • During culture, cells may be imaged without interference from the optical grade silicone
  • Eliminates coverglass overlapping during culture
  • Easily identify "cell side" of coverglass
  • Recommended use temperature is -62 °C to 218 °C

Application

Cell Culture
Immunofluorescence
Microscopy

Legal Information

SecureSlip is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service