Skip to Content
Merck
All Photos(1)

Key Documents

G7153

Sigma-Aldrich

Guanidine hydrochloride

BioXtra, ≥99.5% (titration)

Synonym(s):

Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

product line

BioXtra

Assay

≥99.5% (titration)

impurities

Insoluble matter, passes filter test

ign. residue

≤0.05%

loss

≤0.05% loss on drying, HV, 20°C

pH

4.5-6.0 (6 M in H2O)

mp

180-185 °C (lit.)

solubility

H2O: 6 M, clear, colorless

density

1.3 g/cm3 (lit.)

anion traces

sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%
As: ≤0.0001%
Ba: ≤0.0005%
Bi: ≤0.0005%
Ca: ≤0.001%
Cd: ≤0.0005%
Co: ≤0.0005%
Cr: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Li: ≤0.0005%
Mg: ≤0.0005%
Mn: ≤0.0005%
Mo: ≤0.0005%
Na: ≤0.005%
Ni: ≤0.0005%
Pb: ≤0.0005%
Sr: ≤0.0005%
Zn: ≤0.0005%

absorption

≤0.05 at 280 in H2O at 6 M
≤0.1 at 260 in H2O at 6 M

SMILES string

Cl[H].NC(N)=N

InChI

1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H

InChI key

PJJJBBJSCAKJQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caitriona McKeever et al.
Journal of medicinal chemistry, 56(3), 700-711 (2013-01-11)
Considering the strong DNA minor groove binding observed for our previous series of diaromatic symmetric and asymmetric guanidinium and 2-aminoimidazolinium derivatives, we report now the synthesis of new aminoalkyl derivatives of diaromatic guanidines with potential as DNA minor groove binders
Fackson Mwale et al.
Tissue engineering. Part A, 20(21-22), 2942-2949 (2014-05-03)
Link N is a naturally occurring peptide that can stimulate proteoglycan synthesis in intervertebral disc (IVD) cells. IVD repair can also potentially be enhanced by mesenchymal stem cell (MSC) supplementation to maximize extracellular matrix (ECM) production. In a previous study
Allan Joaquim Lamontanara et al.
Nature communications, 5, 5470-5470 (2014-11-18)
The activity of protein kinases is regulated by multiple molecular mechanisms, and their disruption is a common driver of oncogenesis. A central and almost universal control element of protein kinase activity is the activation loop that utilizes both conformation and
Armando M De Palma et al.
Antimicrobial agents and chemotherapy, 53(5), 1850-1857 (2009-02-25)
A novel compound, TTP-8307, was identified as a potent inhibitor of the replication of several rhino- and enteroviruses. TTP-8307 inhibits viral RNA synthesis in a dose-dependent manner, without affecting polyprotein synthesis and/or processing. Drug-resistant variants of coxsackievirus B3 were all
Sietse T Braakman et al.
Experimental eye research, 130, 87-96 (2014-12-03)
All aqueous humor draining through the conventional outflow pathway must cross the endothelium of Schlemm's canal (SC), likely by passing through micron-sized transendothelial pores. SC pores are non-uniformly distributed along the inner wall endothelium, but it is unclear how the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service