Merck
  • Mutations in the nonstructural protein 3A confer resistance to the novel enterovirus replication inhibitor TTP-8307.
Mutations in the nonstructural protein 3A confer resistance to the novel enterovirus replication inhibitor TTP-8307.
Antimicrobial agents and chemotherapy (2009-02-25)
Armando M De Palma, Hendrik Jan Thibaut, Lonneke van der Linden, Kjerstin Lanke, Ward Heggermont, Stephen Ireland, Robert Andrews, Murty Arimilli, Taleb H Al-Tel, Erik De Clercq, Frank van Kuppeveld, Johan Neyts
ABSTRACT
A novel compound, TTP-8307, was identified as a potent inhibitor of the replication of several rhino- and enteroviruses. TTP-8307 inhibits viral RNA synthesis in a dose-dependent manner, without affecting polyprotein synthesis and/or processing. Drug-resistant variants of coxsackievirus B3 were all shown to carry at least one amino acid mutation in the nonstructural protein 3A. In particular, three mutations located in a nonstructured region preceding the hydrophobic domain (V45A, I54F, and H57Y) appeared to contribute to the drug-resistant phenotype. This region has previously been identified as a hot sport for mutations that resulted in resistance to enviroxime, the sole 3A-targeting enterovirus inhibitor reported thus far. This was corroborated by the fact that TTP-8307 and enviroxime proved cross-resistant. It is hypothesized that TTP-8307 and enviroxime disrupt proper interactions of 3A(B) with other viral or cellular proteins that are required for efficient replication.
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%