Skip to Content
Merck
All Photos(4)

Documents

G4416

Sigma-Aldrich

Anti-GABAA Receptor (α1 subunit) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

antigen ~50 kDa

species reactivity

mouse, rat, human (predicted), bovine (predicted), chicken (predicted)

technique(s)

immunohistochemistry: suitable using rat brain sections
western blot: 1:400-1:1,000 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... GABRA1(2554)
mouse ... Gabra1(14394)
rat ... Gabra1(29705)

General description

GABAA and GABAB receptors differ with regard to their ionic characteristics and pharmacological properties. The GABAA receptor is an ionotropic receptor that forms the GABA gated chloride channel and consists of several heterogeneous subunits with membrane recognition sites for benzodiazapenes.

Immunogen

peptide corresponding to amino acids 28-43 of rat GABA(A) receptor α1 subunit.

Application

Anti-GABAA Receptor (α1 subunit) antibody produced in rabbit is suitable for detection of GABA-A α1 subunit by immunohistochemistry using rat brain sections and by immunoblotting at a working dilution of 1:400-1:1000 using rat brain membranes.

Biochem/physiol Actions

The inhibitory neurotransmitter GABA signals through two distinct types of pre- and postsynaptic receptors, GABAA and GABAB. Both GABA receptors can regulate depression of synaptic transmission and be involved in the inhibition controlling neuronal excitability. α1 is a subtype of GABAA receptor expressed selectively by serotonergic and GABAergic neurons. Serotonergic neurons express strong α3 immunoreactivity but, do not show any α1 immunoreactivity. On the contrary, GABAergic neurons express both α1 and α3 subunits.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 0.05% sodium azide.

Preparation Note

Reconstitute the 0.05 mL vial with 50 μL deionized water. Reconstitute the 0.2 mL vial with 0.2 mL deionized water. After reconstitution, the antibody concentration is ~0.8 mg/mL.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir Jovasevic et al.
Neurobiology of learning and memory, 183, 107459-107459 (2021-05-21)
Alcohol use disorder (AUD) frequently co-occurs with dissociative disorders and disorders with dissociative symptoms, suggesting a common neurobiological basis. It has been proposed that facilitated information processing under the influence of alcohol, resulting in the formation of dissociated memories, might
B Gao et al.
Neuroscience, 54(4), 881-892 (1993-06-01)
GABAA-receptors in the brain display a striking structural heterogeneity, which is based on a multiplicity of diverse subunits. The allocation of GABAA-receptor subtypes to identified neurons is essential for an analysis of the functional significance of receptor heterogeneity. Among GABA-receptive
Diego Armando León Rodríguez et al.
PloS one, 8(6), e68010-e68010 (2013-07-05)
Different models of rodent maternal separation (MS) have been used to investigate long-term neurobiological and behavioral changes, associated with early stress. However, few studies have involved the analysis of sex-related differences in central anxiety modulation. This study investigated whether MS
Hanmeng Zhang et al.
The Journal of physiology, 596(18), 4511-4536 (2018-07-29)
With daily electrophysiological recordings and neurochemical analysis, we uncovered a transient period of synaptic imbalance between enhanced inhibition and suppressed excitation in rat visual cortical neurons from the end of the fourth toward the end of the fifth postnatal weeks.
Qiang-Qiang Xia et al.
Frontiers in cellular neuroscience, 18, 1320784-1320784 (2024-05-28)
Autism Spectrum Disorders (ASDs) are neurodevelopmental disorders (NDDs) in which children display differences in social interaction/communication and repetitive stereotyped behaviors along with variable associated features. Cul3, a gene linked to ASD, encodes CUL3 (CULLIN-3), a protein that serves as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service