Skip to Content
Merck
All Photos(1)

Key Documents

FG0445

SAFC

DMEM - high glucose

With stable glutamine, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

DME, Dulbecco′s Modified Eagle′s Medium - high glucose

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$64.70

$64.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$64.70

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

$64.70


Check Cart for Availability

Request a Bulk Order

Product Name

Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-Ala-L-Gln, sodium pyruvate and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Quality Level

sterility

sterile-filtered
sterile

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

NaHCO3: yes
stable glutamine: yes
glucose: high
phenol red: yes
HEPES: no
sodium pyruvate: yes

shipped in

ambient

storage temp.

2-8°C

General description

This DMEM-Hi glucose medium differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service