Skip to Content
Merck
All Photos(1)

Documents

PEZXMSH01

Millipore

Millicell® EZ Slide

Microscope slide holder for 4 and 8 well units, holds two slides

Synonym(s):

Chamber slides

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.22

material

polystyrene

Quality Level

feature

holds two slides

packaging

pack of 1

manufacturer/tradename

Millicell®

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable
immunocytochemistry: suitable

H

14.7 mm , with device excluding cover
17 mm , with device including cover
9.5 mm

L

128 mm

W

86 mm

color

black

shipped in

ambient

General description

The Millicell® EZ Slide microscope holder fits two Millicell® EZ Slides (4- and 8-well units).

Legal Information

Millicell is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service