Key Documents
EP5427752005
Eppendorf® Fixed-angle rotor
FA-45-24-11 -Kit, incl. rotor lid, aerosol -tight 
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5427752005 is not currently sold in your country. Contact Technical Service
feature
to hold 24 x spin columns
FA-45-24-11 -Kit, incl. rotor lid, aerosol -tight 
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427752005
parameter
13,200 max. RCF (19,090 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)