Skip to Content
Merck
All Photos(1)

Key Documents

EP0030127960

Eppendorf® PCR Sealing Mat

PCR clean, for DWP 96/2000, pkg of 50 ea (5 bags of 10 ea)

Synonym(s):

PCR, Plates, Sealing films

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.15
Product EP0030127960 is not currently sold in your country. Contact Technical Service

sterility

non-sterile

feature

PCR clean
for DWP 96/2000

packaging

pkg of 50 ea (5 bags of 10 ea)

manufacturer/tradename

Eppendorf® 0030127960

technique(s)

PCR: suitable

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service