Skip to Content
Merck
All Photos(1)

Documents

C3731

Greiner CapMat for sealing multiwell plates

96 well CapMat, for sealing 96 well plates, square nap, translucent, non-pierceable

Synonym(s):

multiwell plate cap mats, Greiner CapMats for sealing multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.15

material

ethylene vinyl acetate

sterility

non-sterile

packaging

case of 50 ea (internal packs of 10)

manufacturer/tradename

Greiner 381080

size

96 wells

color

translucent

suitability

suitable for (for sealing 96 well plates)

Looking for similar products? Visit Product Comparison Guide

Chem/Phys Resistance

Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service