EP0030071662
Eppendorf® epT.I.P.S.® PCR Clean
working volume 0.1-5 mL, L 120 mm, violet, pack of 120 ea (5 racks x 24 tips)
Synonym(s):
pipetting tips for PCR
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
violet
sterility
non-sterile
packaging
pack of 120 ea (5 racks x 24 tips)
manufacturer/tradename
Eppendorf® 0030071662
L
120 mm
working volume
0.1-5 mL
Related Categories
General description
Make everyday work easier and more comfortable with the epT.IPS pipette tips (Eppendorf "Totally Integrated Pipetting System"). Manufactured to extremely high standards of quality and purity, these pipette tips are designed to meet all the specific needs of your samples. The epT.IPS require less effort to attach and eject, while completely sealing and perfectly complementing your Eppendorf pipettes, with which they form a fully integrated pipetting system that offers maximum precision, accuracy and reproducibility.
The Eppendorf pipette tips with their ideal wetting properties and optimal transparency are made exclusively from high-purity plastics. Thanks to their universal cone, the Eppendorf tips can also be used with pipettes from other manufacturers.
The Eppendorf pipette tips with their ideal wetting properties and optimal transparency are made exclusively from high-purity plastics. Thanks to their universal cone, the Eppendorf tips can also be used with pipettes from other manufacturers.
Application
PCR
Features and Benefits
Long, narrow tips reach the base of narrow and high vessels without touching the top of the vessel edge
Modern slim rack design with reclosable lid
Improved sustainability through 35 % lower polypropylene consumption per rack
Modern slim rack design with reclosable lid
Improved sustainability through 35 % lower polypropylene consumption per rack
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service