Skip to Content
Merck
All Photos(1)

Documents

EMU052501

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Prkaa2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGTGGATCGCCAAATTATGCAGCACCTGAGGTCATCTCAGGAAGGCTGTATGCAGGTCCCGAGGTCGATATCTGGAGCTGTGGTGTCATCCTGTATGCCCTTCTCTGTGGCACCCTCCCTTTCGATGATGAGCACGTGCCTACGCTCTTCAAGAAGATCCGAGGGGGTGTGTTTTACATCCCAGACTATCTCAACCGTTCTGTCGCCACTCTGCTGATGCACATGCTCCAGGTGGACCCCCTGAAGCGAGCGACTATCAAAGACATACGAGAACATGAATGGTTTAAACAGGATTTGCCCAGCTACCTATTTCCTGAAGACCCCTCCTACGATGCGAATGTCATTGTCGATGAGGCTGTGAAGGAAGTCTGTGAGAAATTCGAGTGTACAGAGTCAGAAGTGATGAATAGTCTGTATAGTGGTGACCCTCAAGACCAGCTTGCAGTGGCTTAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sreevidya Santha et al.
The Journal of biological chemistry, 290(36), 21865-21875 (2015-07-23)
Prostate cancer (PCa) is one of the most frequently diagnosed cancers in men with limited treatment options for the hormone-resistant forms. Development of novel therapeutic options is critically needed to target advanced forms. Here we demonstrate that combinatorial treatment with
Sravanth K Hindupur et al.
Breast cancer research : BCR, 16(4), 420-420 (2014-08-07)
Matrix detachment triggers anoikis, a form of apoptosis, in most normal epithelial cells, while acquisition of anoikis resistance is a prime requisite for solid tumor growth. Of note, recent studies have revealed that a small population of normal human mammary
Xia Cao et al.
Hypertension research : official journal of the Japanese Society of Hypertension, 37(9), 803-810 (2014-06-27)
The purpose of this study was to determine the effects of resveratrol (RSV) and the molecular mechanisms by which it regulates vascular smooth muscle contraction and blood pressure in mice. In cultured human vascular smooth muscle cells (VSMCs), we found
Teraneh Z Jhaveri et al.
Oncotarget, 6(17), 14754-14765 (2015-07-06)
AMP-activated Protein Kinase (AMPK) activity retards growth of many types of cancers. Investigating effects of AMPK activation on breast cancer cell signaling and survival, we found that breast cancer cell lines with amplification and over-expression of HER2 or EGFR are
Chiara Zucal et al.
BMC cancer, 15, 855-855 (2015-11-07)
Nicotinamide phosphoribosyltransferase (NAMPT), the rate-limiting enzyme in NAD(+) biosynthesis from nicotinamide, is one of the major factors regulating cancer cells metabolism and is considered a promising target for treating cancer. The prototypical NAMPT inhibitor FK866 effectively lowers NAD(+) levels in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service