Skip to Content
Merck
All Photos(1)

Key Documents

EHU084871

Sigma-Aldrich

MISSION® esiRNA

targeting human JAK1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on16 April 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on16 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAAATGCCTGGCTCCTATTCGAGACCCCAAGACCGAGCAGGATGGACATGATATTGAGAACGAGTGTCTAGGGATGGCTGTCCTGGCCATCTCACACTATGCCATGATGAAGAAGATGCAGTTGCCAGAACTGCCCAAGGACATCAGCTACAAGCGATATATTCCAGAAACATTGAATAAGTCCATCAGACAGAGGAACCTTCTCACCAGGATGCGGATAAATAATGTTTTCAAGGATTTCCTAAAGGAATTTAACAACAAGACCATTTGTGACAGCAGCGTGTCCACGCATGACCTGAAGGTGAAATACTTGGCTACCTTGGAAACTTTGACAAAACATTACGGTGCTGAAATATTTGAGACTTCCATGTTACTGATTTCATCAGAAAATGAGATGAATTGGTTTCATTCGAATGACGGTGGAAACGTTCTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matija Hedl et al.
Journal of immunology (Baltimore, Md. : 1950), 197(9), 3695-3704 (2016-09-25)
JAK2 genetic variants are associated with inflammatory bowel disease (IBD) and JAK inhibitors are being evaluated for therapy targeting immune-mediated diseases, including IBD. As JAK pathway-mediated cytokine regulation varies across cell types and stimulation conditions, we examined how JAK signaling
Elisabeth Losdyck et al.
The Journal of biological chemistry, 290(48), 29022-29034 (2015-10-09)
JAK1 and JAK3 are recurrently mutated in acute lymphoblastic leukemia. These tyrosine kinases associate with heterodimeric cytokine receptors such as IL-7 receptor or IL-9 receptor, in which JAK1 is appended to the specific chain, and JAK3 is appended to the
Li-Chuan Chan et al.
The Journal of clinical investigation, 129(8), 3324-3338 (2019-07-16)
Glycosylation of immune receptors and ligands, such as T cell receptor and coinhibitory molecules, regulates immune signaling activation and immune surveillance. However, how oncogenic signaling initiates glycosylation of coinhibitory molecules to induce immunosuppression remains unclear. Here we show that IL-6-activated
Han Liu et al.
Oncotarget, 8(24), 38113-38135 (2017-05-13)
Human colon cancers express higher levels of NADPH oxidase 1 [NOX1] than adjacent normal epithelium. It has been suggested that reactive oxygen species [ROS] derived from NOX1 contribute to DNA damage and neoplastic transformation in the colon, particularly during chronic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service