Skip to Content
Merck
All Photos(3)

Documents

B4387

Sigma-Aldrich

5-Bromo-3-indolyl β-D-galactopyranoside

≥98%, powder

Synonym(s):

Bluo-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16BrNO6
CAS Number:
Molecular Weight:
374.18
Beilstein:
1550140
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

5-Bromo-3-indolyl β-D-galactopyranoside, ≥98%

Assay

≥98%

form

powder

solubility

DMF: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)cc23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H16BrNO6/c15-6-1-2-8-7(3-6)9(4-16-8)21-14-13(20)12(19)11(18)10(5-17)22-14/h1-4,10-14,16-20H,5H2/t10-,11+,12+,13-,14-/m1/s1

InChI key

LINMATFDVHBYOS-MBJXGIAVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chromogenic substrate suitable for identification of lac+ bacterial colonies. An alternative to 5-bromo-4-chloro-3-indolyl β-D-galactopyranoside (X-Gal), producing a darker blue color.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Kodaka et al.
Journal of clinical microbiology, 33(1), 199-201 (1995-01-01)
A new medium containing 5-bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt (Glu agar) for Escherichia coli and a new medium containing 5-bromo-3-indolyl-beta-D-galactoside (Gal agar) for beta-galactosidase-positive members of the family Enterobacteriaceae were compared with MacConkey agar in a diagnostic trial with 3,562 urine specimens.
A N Markarian et al.
Bioorganicheskaia khimiia, 13(2), 263-265 (1987-02-01)
A simple and convenient technique has been developed for detection of beta-galactosidase from E. coli on nitrocellulose sheets using a mixture of 5-bromoindol-3-yl-beta-D-galactopyranoside and nitro blue tetrazolium, which enables rapid detection of fmole (10(-15) mole) quantities of the enzyme at
Tushar Gupta et al.
Journal of virology, 89(9), 5124-5133 (2015-02-27)
The E2F family of transcription factors, broadly divided into activator and repressor E2Fs, regulates cell cycle genes. Current models indicate that activator E2Fs are necessary for cell cycle progression and tumorigenesis and are also required to mediate transformation induced by
Hongjun Chen et al.
Journal of virology, 88(17), 10013-10025 (2014-06-20)
Vaccination is the first line of defense against influenza virus infection, yet influenza vaccine production methods are slow, antiquated, and expensive as a means to effectively reduce the virus burden during epidemic or pandemic periods. There is a great need

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service