Skip to Content
Merck
All Photos(1)

Key Documents

EHU066711

Sigma-Aldrich

MISSION® esiRNA

targeting human BMX

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on02 June 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on02 June 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAGGCCCAGACTATGATGAAACTCAGCCATCCCAAGCTGGTTAAATTCTATGGAGTGTGTTCAAAGGAATACCCCATATACATAGTGACTGAATATATAAGCAATGGCTGCTTGCTGAATTACCTGAGGAGTCACGGAAAAGGACTTGAACCTTCCCAGCTCTTAGAAATGTGCTACGATGTCTGTGAAGGCATGGCCTTCTTGGAGAGTCACCAATTCATACACCGGGACTTGGCTGCTCGTAACTGCTTGGTGGACAGAGATCTCTGTGTGAAAGTATCTGACTTTGGAATGACAAGGTATGTTCTTGATGACCAGTATGTCAGTTCAGTCGGAACAAAGTTTCCAGTCAAGTGGTCAGCTCCAGAGGTGTTTCATTACTTCAAATACAGCAGCAAGTCAGACGTATGGGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... BMX(660) , BMX(660)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Wei et al.
American journal of cancer research, 7(3), 628-646 (2017-04-13)
miR-495 serves as an oncogenic miRNA or a tumor suppressor in different types of cancer. However, its role in the drug resistance of small cell lung cancer (SCLC) remains unidentified. In this study, we investigated whether miR-495 regulates the chemoresistance
Lesley A Mathews et al.
Molecular cancer, 9, 267-267 (2010-10-12)
Recently, much attention has been focused on gaining a better understanding of the different populations of cells within a tumor and their contribution to cancer progression. One of the most commonly used methods to isolate a more aggressive sub-population of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service