Skip to Content
Merck
All Photos(1)

Documents

EHU043401

Sigma-Aldrich

MISSION® esiRNA

targeting human LRP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTGGATGTGGATGTGTCCTCTGGCTTTATTTATTGGTGTGATTTTAGCAGCTCAGTGGCATCTGATAATGCGATCCGTAGAATTAAACCAGATGGATCTTCTCTGATGAACATTGTGACACATGGAATAGGAGAAAATGGAGTCCGGGGTATTGCAGTGGATTGGGTAGCAGGAAATCTTTATTTCACCAATGCCTTTGTTTCTGAAACACTGATAGAAGTTCTGCGGATCAATACTACTTACCGCCGTGTTCTTCTTAAAGTCACAGTGGACATGCCTAGGCATATTGTTGTAGATCCCAAGAACAGATACCTCTTCTGGGCTGACTATGGGCAGAGACCAAAGATTGAGCGTTCTTTCCTTGACTGTACCAATCGAACAGTGCTTGTGTCAGAGGGCATTGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Gao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(6), 7684-7693 (2019-03-21)
Osteoblast differentiation of human mesenchymal stem cells (hMSCs) is stimulated by 1α,25-dihydroxycholecalciferol [1α,25(OH)2D3] and 25-hydroxycholecalciferol [25(OH)D3]; the latter's effects require intracellular hydroxylation to 1α,25(OH)2D3. Thus, hMSCs are both a source of and target for 1α,25(OH)2D3. Megalin is a transmembrane receptor
Aurélien Briens et al.
Cell discovery, 3, 17001-17001 (2017-04-19)
Plasminogen activation is involved in many processes within the central nervous system, including synaptic plasticity, neuroinflammation and neurodegeneration. However, the mechanisms that regulate plasminogen activation in the brain still remain unknown. Here we demonstrate that astrocytes participate in this regulation
Rohit Upadhyay et al.
JCI insight, 5(14) (2020-06-17)
Free light chains (FLCs) induce inflammatory pathways in proximal tubule cells (PTCs). The role of TLRs in these responses is unknown. Here we present findings on the role of TLRs in FLC-induced PTC injury. We exposed human kidney PTC cultures
Kimberly R Long et al.
American journal of physiology. Renal physiology, 318(3), F851-F859 (2020-02-19)
Albuminuria is frequently associated with proximal tubule (PT) cytotoxicity that can feed back to cause glomerular damage and exacerbate kidney disease. PT cells express megalin and cubilin receptors that bind to and internalize albumin over a broad concentration range. How
Jeanne L Theis et al.
eLife, 9 (2020-10-03)
Congenital heart diseases (CHDs), including hypoplastic left heart syndrome (HLHS), are genetically complex and poorly understood. Here, a multidisciplinary platform was established to functionally evaluate novel CHD gene candidates, based on whole-genome and iPSC RNA sequencing of a HLHS family-trio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service