Merck
  • Astrocytes regulate the balance between plasminogen activation and plasmin clearance via cell-surface actin.
Astrocytes regulate the balance between plasminogen activation and plasmin clearance via cell-surface actin.
Cell discovery (2017-04-19)
Aurélien Briens, Isabelle Bardou, Héloïse Lebas, Lindsey A Miles, Robert J Parmer, Denis Vivien, Fabian Docagne
ABSTRACT
Plasminogen activation is involved in many processes within the central nervous system, including synaptic plasticity, neuroinflammation and neurodegeneration. However, the mechanisms that regulate plasminogen activation in the brain still remain unknown. Here we demonstrate that astrocytes participate in this regulation by two mechanisms. First, the astrocyte plasma membrane serves as a surface for plasminogen activation by tissue-type plasminogen activator. This activation triggers downstream plasmin-dependent processes with important impacts in brain health and disease, such as fibrinolysis and brain-derived neurotrophic factor conversion. Second, astrocytes take up plasminogen and plasmin in a regulated manner through a novel mechanism involving endocytosis mediated by cell-surface actin and triggered by extracellular plasmin activity at the surface of astrocytes. Following endocytosis, plasminogen and plasmin are targeted to lysosomes for degradation. Thus, cell-surface actin acts as a sensor of plasmin activity to induce a negative feedback through plasmin endocytosis. This study provides evidence that astrocytes control the balance between plasmin formation and plasmin elimination in the brain parenchyma.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Dynasore hydrate
Sigma-Aldrich
MISSION® esiRNA, targeting human LRP2
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution