Skip to Content
Merck
All Photos(3)

Documents

E7781

Sigma-Aldrich

Erastin

≥98% (HPLC), powder, antitumor agent

Synonym(s):

2-[1-[4-[2-(4-Chlorophenoxy)acetyl]-1-piperazinyl]ethyl]-3-(2-ethoxyphenyl)-4(3H)-Quinazolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H31ClN4O4
CAS Number:
Molecular Weight:
547.04
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Erastin, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CCOc1ccccc1N2C(=O)c3ccccc3N=C2C(C)N4CCN(CC4)C(=O)COc5ccc(Cl)cc5

InChI

1S/C30H31ClN4O4/c1-3-38-27-11-7-6-10-26(27)35-29(32-25-9-5-4-8-24(25)30(35)37)21(2)33-16-18-34(19-17-33)28(36)20-39-23-14-12-22(31)13-15-23/h4-15,21H,3,16-20H2,1-2H3

InChI key

BKQFRNYHFIQEKN-UHFFFAOYSA-N

Gene Information

human ... hRas(3265)
mouse ... hRas(15461)
rat ... hRas(293621)

General description

Erastin is a cell-permeable piperazinyl-quinazolinone. It interacts with antiporter system Xc-.

Application

Erastin has been used:
  • as a positive control for inducing ferroptosis in hepatic stellate cell (HSC)
  • to induce ferroptosis and in transferrin internalization assay of human fibrosarcoma HT1080 cells
  • to induce ferroptosis of muscle-derived cell lines

Biochem/physiol Actions

Erastin is an antitumor agent selective for tumor cells bearing oncogenic RAS (i.e. HRAS, KRAS). Erastin produces ferroptosis, a non-apoptotic tumor cell death, by altering mitochondrial voltage-dependent anion channel (VDAC) gating allowing cations to enter mitochondria and leading to release of oxidative species causing oxidative cell death.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cell growth potential drives ferroptosis susceptibility in rhabdomyosarcoma and myoblast cell lines
Codenotti S, et al.
Journal of Cancer Research and Clinical Oncology, 144(9), 1717-1730 (2018)
Sonam Dolma et al.
Cancer cell, 3(3), 285-296 (2003-04-05)
We used synthetic lethal high-throughput screening to interrogate 23,550 compounds for their ability to kill engineered tumorigenic cells but not their isogenic normal cell counterparts. We identified known and novel compounds with genotype-selective activity, including doxorubicin, daunorubicin, mitoxantrone, camptothecin, sangivamycin
The ferroptosis inducer erastin promotes proliferation and differentiation in human peripheral blood mononuclear cells
Wang D, et al.
Biochemical and Biophysical Research Communications, 503(3), 1689-1695 (2018)
Jiao Wu et al.
Nature, 572(7769), 402-406 (2019-07-26)
Ferroptosis, a cell death process driven by cellular metabolism and iron-dependent lipid peroxidation, has been implicated in diseases such as ischaemic organ damage and cancer1,2. The enzyme glutathione peroxidase 4 (GPX4) is a central regulator of ferroptosis, and protects cells
Antonio Bruni et al.
Cell death & disease, 9(6), 595-595 (2018-05-24)
Human islet transplantation has been hampered by donor cell death associated with the islet preparation procedure before transplantation. Regulated necrosis pathways are biochemically and morphologically distinct from apoptosis. Recently, ferroptosis was identified as a non-apoptotic form of iron-dependent regulated necrosis

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Apoptosis regulation involves multiple pathways and molecules for cellular homeostasis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service