Skip to Content
Merck
All Photos(1)

Key Documents

E2906

Sigma-Aldrich

Anti-dEGF Receptor, Extracellular Domain antibody, Mouse monoclonal

clone C-273, purified from hybridoma cell culture

Synonym(s):

Anti-Drosophila Epidermal Growth Factor Receptor, Anti-dEGFR

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$811.00

$811.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$811.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$811.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

C-273, monoclonal

form

buffered aqueous solution

species reactivity

Drosophila

concentration

~1.2 mg/mL

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: 0.25-0.5 μg/mL using recombinant protein representing amino acids 299-359 of the Drosophila EGFR extracellular domain

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Drosophila melanogaster ... Egfr(37455)

General description

Anti-dEGF Receptor, Extracellular Domain antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma C-273 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with a recombinant protein encoding at the extracellular portion of Drosophila epidermal growth factor receptor (EGFR). Drosophila EGF receptor, also known as Torpedo or DER has four ligands: Gurken, Spitz (the principle ligand), Vein and Argos. Two accessory proteins modulate its signalling: Rhomboid and Star.

Application

Suitable for the following:
  • Immunohistochemistry
  • Indirect ELISA
  • Western blotting at a concentration of 0.25-0.5μg/mL using recombinant protein representing amino acids 299-359 of the Drosophila EGFR extracellular domain

Biochem/physiol Actions

Drosophila epidermal growth factor receptor (EGFR) is involved in many developmental processes such as: egg polarity, cell identity in the ventral ectoderm, neurogenesis, development of the Malpighian tubules, and larval eye and wing development. The downstream signalling molecules that are activated by the Drosophila EGF receptor include: Src homology (Shc), DRK (a homolog of mammalian Grb2), a guanine nucleotide exchange factor (SOS) activated by DRK, and downstream targets including orthologs of Ras, Raf, and Rolled (MAP Kinase).
The epidermal growth factor receptor (EGFR) binds to its cognate ligands that lead to autophosphorylation of receptor tyrosine kinase and subsequent activation of signal transduction pathways that are involved in regulating cellular proliferation, differentiation and survival. EGFR is overexpressed in tumor cell lines and is associated with poor prognosis leading to decreased survival. Its activation also plays a role in resistance to chemotherapy and radiation treatment in tumor cells. EGFR tyrosine kinase inhibitors (TKIs) may act as a new treatment option for advanced lung adenocarcinoma. Its ligands function in the Drosophila CNS and maintains glial survival. Drosophila EGF receptor plays a critical role during the development of Drosophila retina.

Physical form

Solution in 0.01 M phosphate buffered saline containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulating the dynamics of EGF receptor signaling in space and time
Shilo BZ
Development, 132(18), 4017-4027 (2005)
Roy S Herbst
International journal of radiation oncology, biology, physics, 59(2 Suppl), 21-26 (2004-05-15)
The epidermal growth factor receptor (EGFR) is a transmembrane glycoprotein that constitutes one of four members of the erbB family of tyrosine kinase receptors. Binding of EGFR to its cognate ligands leads to autophosphorylation of receptor tyrosine kinase and subsequent
Rebecca P Kim-Yip et al.
Development (Cambridge, England), 145(23) (2018-11-06)
Adult stem cell niche boundaries must be precisely maintained to facilitate the segregation of stem cell and daughter cell fates. However, the mechanisms that govern this process in epithelial tissues are not fully understood. In this study, we investigated the
Josefa Steinhauer et al.
Journal of cell science, 126(Pt 19), 4469-4478 (2013-08-02)
Epidermal growth factor receptor (EGFR) ligands undergo a complex series of processing events during their maturation to active signaling proteins. Like its mammalian homologs, the predominant Drosophila EGFR ligand Spitz is produced as a transmembrane pro-protein. In the secretory pathway
Nicole T Revaitis et al.
Development (Cambridge, England), 147(15) (2020-07-19)
To bridge the gap between qualitative and quantitative analyses of the epidermal growth factor receptor (EGFR) in tissues, we generated an sfGFP-tagged EGF receptor (EGFR-sfGFP) in Drosophila The homozygous fly appears similar to wild type with EGFR expression and activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service