Skip to Content
Merck
All Photos(14)

Key Documents

ZRB1220

Sigma-Aldrich

Anti-GAP43 Antibody, clone 1D6 , ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Axonal membrane protein GAP-43, Growth-associated protein 43, Neural phosphoprotein B-50, Neuromodulin, pp46

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
$303.00
4 X 25 μL
$625.00

$303.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
$303.00
4 X 25 μL
$625.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$303.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D6, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 24.8 kDa
observed mol wt ~47 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... GAP43(2596)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D6 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects human Growth-Associated Protein 43 (GAP43).

Immunogen

GST/His-tagged recombinant fragment corresponding to the first 126 amino acids from the N-terminal half of human Growth-Associated Protein 43 (GAP43).

Application

Anti-GAP43, clone 1D6 ZooMAb, Cat. No. ZRB1220, is a highly specific rabbit recombinant monoclonal antibody that targets Growth-associated protein 43 (GAP43) and has been tested for use Immunocytochemistry, Immunohistochemistry (Paraffin). And Western Blotting.
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representiatve lot detected GAP43 in human and mouse cerebral cortex tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representiatve lot detected GAP43 in SH-SY5Y cells.

Note: Actual optimal working dilutions must be determined by end user as specimens and experimental conditions may vary.

Target description

Neuromodulin (UniProt: P17677; also known as Axonal membrane protein GAP-43, Growth-associated protein 43, Neural phosphoprotein B-50, pp46) is encoded by the GAP43 gene (Gene ID: 2596) in human. GAP43 is a peripheral membrane protein that is associated with nerve growth. It serves as a common marker of differentiating neurons. It is a major component of the motile "growth cones" that form the tips of elongating axons and plays a role in axonal and dendritic filopodia induction. It is expressed at high levels in neuronal growth cones during development and axonal regeneration. However, it is not fond in dendritic cones. In adult brain, GAP43 is found in high concentration in presynaptic areas where memory formation is thought to occur, such as frontal cortex, limbic system and hippocampus. GAP43 contains an N-terminal lipid binding IQ domain (aa 31-60) that also binds calmodulin. GAP43 is reported to bind calmodulin at low calcium levels and then releases it when calcium levels increase indicating that calmodulin may serve as a negative regulator of GAP43 during periods of low neuronal activity. Protein kinase C-mediated phosphorylation serine 41 is reported to inhibit this interaction and stabilizes actin filaments and promotes growth cone extension. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.(Ref.: Neve, RL et al., (1998). J. Neurosci. 18(19); 7757-7767).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service