Skip to Content
Merck
All Photos(2)

Key Documents

D3070

Sigma-Aldrich

Anti-DCDC2 (C-terminal) antibody produced in rabbit

enhanced validation

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DCDC2A, Anti-RU2, Anti-RU2S

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$618.00

$618.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$618.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$618.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa
antigen ~70 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.5 mg/mL

technique(s)

western blot: 0.5-1 μg/mL using HEK-293T cell lysate expressing human DCDC2

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCDC2(51473)

General description

Doublecortin domain-containing protein 2 (DCDC2) belongs to the doublecortin (DCX) superfamily and contains two doublecortin domains. DCDC2 is expressed in the fetal and adult central nervous system (CNS). DCDC2 gene is mapped to human chromosome 6p22.3.

Specificity

Anti-DCDC2 (C-terminal) specifically recognizes human DCDC2.

Application

Anti-DCDC2 antibody produced in rabbit is suitable for western blot analysis at a working concentration of 0.5-1μg/mL using a HEK-293T cell lysate expressing human DCDC2.

Biochem/physiol Actions

DCDC2 is one of the dyslexia susceptibility genes involved in neuronal migration and other developmental processes. The DCX domain of DCDC2 binds tubulin and enhances microtubule polymerization. Several SNPs within the DCDC2 gene have been associated with dyslexia. It modulates neuronal development in the brain.
Mutations in the DCDC2 gene is implicated in non-syndromic recessive deafness DFNB66 neonatal sclerosing cholangitis (NSC) and renal-hepatic ciliopathy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orly Reiner et al.
BMC genomics, 7, 188-188 (2006-07-28)
Doublecortin (DCX) domains serve as protein-interaction platforms. Mutations in members of this protein superfamily are linked to several genetic diseases. Mutations in the human DCX gene result in abnormal neuronal migration, epilepsy, and mental retardation; mutations in RP1 are associated
Yuxiang Lin et al.
Clinics and research in hepatology and gastroenterology, 44(5), e103-e108 (2020-03-25)
Neonatal sclerosing cholangitis (NSC) is a severe cholestatic liver disease, which often develops into end-stage liver disease in childhood and requires liver transplantation. Mutations in CLDN1 and DCDC2 are confirmed to be the main pathogenic mechanism of NSC. Whole exon
Lauren M McGrath et al.
Trends in molecular medicine, 12(7), 333-341 (2006-06-20)
Four genes have recently been proposed as candidates for dyslexia: dyslexia susceptibility 1 candidate 1 (DYX1C1), roundabout Drosophila homolog 1 (ROBO1), doublecortin domain-containing protein 2 (DCDC2) and KIAA0319. Each gene is implicated in global brain-development processes such as neural migration
Johannes Schumacher et al.
American journal of human genetics, 78(1), 52-62 (2005-12-31)
We searched for linkage disequilibrium (LD) in 137 triads with dyslexia, using markers that span the most-replicated dyslexia susceptibility region on 6p21-p22, and found association between the disease and markers within the VMP/DCDC2/KAAG1 locus. Detailed refinement of the LD region
Markus Schueler et al.
American journal of human genetics, 96(1), 81-92 (2015-01-06)
Nephronophthisis-related ciliopathies (NPHP-RC) are recessive diseases characterized by renal dysplasia or degeneration. We here identify mutations of DCDC2 as causing a renal-hepatic ciliopathy. DCDC2 localizes to the ciliary axoneme and to mitotic spindle fibers in a cell-cycle-dependent manner. Knockdown of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service