Skip to Content
Merck
All Photos(2)

Key Documents

D2155

Sigma-Aldrich

N-Dansyl-D-Ala-Gly-p-nitro-Phe-Gly

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$229.00

$229.00


Check Cart for Availability


Select a Size

Change View
25 MG
$229.00

About This Item

Empirical Formula (Hill Notation):
C28H32N6O9S
CAS Number:
Molecular Weight:
628.65
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

$229.00


Check Cart for Availability

Quality Level

Assay

≥95% (HPLC)

form

powder

solubility

methanol: 50 mg

storage temp.

−20°C

SMILES string

CC(NS(=O)(=O)c1cccc2c(cccc12)N(C)C)C(=O)NCC(=O)NC(Cc3ccc(cc3)N(=O)=O)C(=O)NCC(O)=O

InChI

1S/C28H32N6O9S/c1-17(32-44(42,43)24-9-5-6-20-21(24)7-4-8-23(20)33(2)3)27(38)29-15-25(35)31-22(28(39)30-16-26(36)37)14-18-10-12-19(13-11-18)34(40)41/h4-13,17,22,32H,14-16H2,1-3H3,(H,29,38)(H,30,39)(H,31,35)(H,36,37)

InChI key

MIHGGGACSSHXPY-UHFFFAOYSA-N

Related Categories

Substrates

Selective fluorogenic substrate for enkephalinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hitoshi Nakagawa et al.
Hypertension (Dallas, Tex. : 1979), 77(6), 1931-1939 (2021-04-13)
[Figure: see text].
Rishi K Trivedi et al.
Journal of the American Heart Association, 7(5) (2018-03-05)
There is a paucity of data about the mechanisms by which sacubitril/valsartan (also known as LCZ696) improves outcomes in patients with heart failure. Specifically, the effects of sacubitril/valsartan on vascular function and NO bioavailability have not been investigated. We hypothesized
D Florentin et al.
Analytical biochemistry, 141(1), 62-69 (1984-08-15)
A fluorogenic peptide, dansyl-D-Ala-Gly-Phe(pNO2)-Gly (DAGNPG), was synthesized as a selective substrate for the neutral metalloendopeptidase (EC 3.4.24.11) involved in enkephalin metabolism. This enzyme, designated "enkephalinase," cleaves the Gly-Phe(pNO2) peptide bond of DAGNPG (V = 0.65 mumol/mg protein/min and Km =

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service