Skip to Content
Merck
All Photos(1)

Documents

SRP6450

Sigma-Aldrich

Neprilysin/CD10 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CALLA, DKFZp686O16152, MGC126681, MGC126707, MME, NEP, SFE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

calculated mol wt 80 kDa
observed mol wt 90-100 kDa (DTT-reduced. Tyr 52 is the predicted N-terminal .)

packaging

pkg of 10 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MME(4311)

General description

Cluster of differentiation 10 (CD10) is also known as membrane metallo-endopeptidase, neutral endopeptidase (NEP), neprilysin and common acute lymphoblastic leukemia antigen (CALLA). It is a 90-110kDa type II transmembrane glycoprotein normally expressed by a variety of tissues, including epithelial cells of the prostate, kidney, intestine, endometrium, adrenal glands and lung. The protein has a cytoplasmic amino-terminal domain, a transmembrane hydrophobic domain, and an extracellular domain which possesses catalytic activity. The gene encoding this protein is localized on human chromosome 3q25.2.

Biochem/physiol Actions

Cluster of differentiation 10 (CD10) is a zinc-dependent metalloprotease enzyme that cleaves peptide bonds on the amino side of hydrophobic residues and inactivates a variety of physiologically active secreted peptides. It is thought to be the rate-limiting degrading enzyme of amyloid-β peptide (Aβ) whose abnormal misfolding and aggregation in neural tissue has been implicated in the development of Alzheimer′s disease (AD). CD10 is also identified as the common acute lymphoblastic leukemia antigen (CALLA) present on leukemic cells of pre-B phenotype, and thus serves as the most important biomarker in the diagnosis of human acute lymphocytic leukemia (ALL).

Physical form

Lyophilized from 0.22 μm filtered solution in 50 mM tris, 150 mM NaCl, pH 8.0 Normally Mannitol or Trehalose is added as protectants before lyophilization.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile PBS, pH 7.4 to a concentration of 50 μg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 month. For extended storage, it is recommended to store at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari van de Vyver et al.
Stem cells and development, 30(23), 1141-1152 (2021-06-17)
Monitoring wound progression over time is a critical aspect for studies focused on in-depth molecular analysis or on evaluating the efficacy of potential novel therapies. Histopathological analysis of wound biopsies can provide significant insight into healing dynamics, yet there is
Role of B Cell Development Marker CD10 in Cancer Progression and Prognosis.
Mishra D
Molecular Biology International, 4328697-4328697 (2016)
Kiara Boodhoo et al.
Journal of molecular endocrinology, 66(2), 141-155 (2020-12-23)
Chronic wounds are a serious and debilitating complication of diabetes. A better understanding of the dysregulated healing responses following injury will provide insight into the optimal time frame for therapeutic intervention. In this study, a direct comparison was done between
Luca Dellafiora et al.
Toxins, 12(4) (2020-04-23)
Ochratoxin A (OTA), a mycotoxin that is of utmost concern in food and feed safety, is produced by fungal species that mainly belong to the Aspergillus and Penicillium genera. The development of mitigation strategies to reduce OTA content along the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service