Skip to Content
Merck
All Photos(1)

Documents

D1819

Sigma-Aldrich

Anti-DYRK1A (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Dual-specificity tyrosine-phosphorylation-regulated kinase 1A, Anti-MNB protein kinase, Anti-MNB/MNBH, Anti-Minibrain Drosophila homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~86 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using rat brain embryonic extract
western blot: 2-4 μg/mL using rat brain postnatal extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DYRK1A(1859)
mouse ... Dyrk1a(13548)
rat ... Dyrk1a(25255)

General description

Dual-specificity tyrosine-phosphorylated regulated kinase 1A (DYRK1A), also known as minibrain/Mnb is a member of a growing family of Ser/Thr protein kinases termed dual-specificity tyrosine phosphorylation-regulated kinases (DYRKs). DYRKs also possess autophosphorylation activity on tyrosine residues, located on the YXY motif of the activation loop of the catalytic domain.
The human and rodent DYRK1A are ubiquitously expressed in adult and fetal tissue with high expression in the brain and heart during development.

Application

Anti-DYRK1A (C-terminal) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

DYRK1A is encoded by a gene located within the Down syndrome (DS) critical region on human chromosome 21 and its expression is found to be elevated in individuals with DS. It might be one of the genes involved in some of the neurological abnormalities observed in DS. It phosphorylates several substrates including transcription factor FKHR, NFAT, STAT3, microtubule-associated protein Tau, glycogen synthase and c-AMP-response element-binding protein.
DYRK1A phosphorylates several substrates including transcription factor FKHR, NFAT, STAT3, microtubule-associated protein Tau, glycogen synthase and c-AMP-response element-binding protein.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DYRK1A-haploinsufficiency in mice causes autistic-like features and febrile seizures
Raveau M, et al.
Neurobiology of Disease, 110, 180-191 (2018)
Emerging Roles of DYRK Kinases in Embryogenesis and Hedgehog Pathway Control
Singh R, et al.
Journal of developmental biology, 5(4) (2017)
Rodolphe Dard et al.
Genes, 12(11) (2021-11-28)
Down syndrome (DS) is the most common chromosomal disorder. It is responsible for intellectual disability (ID) and several medical conditions. Although men with DS are thought to be infertile, some spontaneous paternities have been reported. The few studies of the
W J Song et al.
Genomics, 38(3), 331-339 (1996-12-15)
The presence of an extra copy of human chromosome 21 (trisomy 21), especially region 21q22.2, causes many phenotypes in Down syndrome, including mental retardation. To study genes potentially responsible for some of these phenotypes, we cloned a human candidate gene
Noriko Murakami et al.
The Journal of biological chemistry, 281(33), 23712-23724 (2006-05-31)
Minibrain kinase/dual-specificity tyrosine phosphorylation-regulated kinase (Mnb/Dyrk1A) is a proline-directed serine/threonine kinase encoded in the Down syndrome critical region of human chromosome 21. This kinase has been shown to phosphorylate dynamin 1 and synaptojanin 1. Here we report that amphiphysin I

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service