Skip to Content
Merck
All Photos(5)

Documents

HPA015323

Sigma-Aldrich

Anti-DYRK1A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dual specificity YAK1-related kinase, Anti-Dual specificity tyrosine-phosphorylation-regulated kinase 1A, Anti-HP86, Anti-MNBH, Anti-Protein kinase minibrain homolog, Anti-hMNB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VRQQFPAPLGWSGTEAPTQVTVETHPVQETTFHVAPQQNALHHHHGNSSHHHHHHHHHHHHHGQQALGNRTRPRVYNSPTNSSSTQDSMEVGHSHHSMTSLSSSTTSSSTSSSSTG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DYRK1A(1859)

General description

DYRK1A (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A) is a highly conserved kinase, which is a member of the CMGC family of protein kinases. It is a serine/threonine kinase, which is a Drosophila minibrain gene homolog. This gene is localized to human chromosome 21, and is expressed in both nucleus and cytoplasm. It has a ubiquitous expression pattern, and shows predominant expression in hippocampus, cerebellum and olfactory bulb.

Immunogen

Dual specificity tyrosine-phosphorylation-regulated kinase 1A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

DYRK1A (dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1A) is involved in the homeostasis of tissues, and in human development. This gene is up-regulated in Down syndrome (DS) patients, which is linked with changes in motor functions, osteoporosis, aberrations in retina etc. Mutations resulting in truncated protein are linked to retardation in general growth and severe primary microcephaly. Mutations in this gene are also associated with deregulation of pancreatic function, and progression of tumor. This gene is down-regulated in Alzheimer′s disease, and its plasma levels might be diagnostic marker for the same. It acts as a tumor suppressor in acute myeloid leukemia (AML), and its expression is suppressed in the same. It suppresses the proliferation of AML cells, by degrading c-Myc, and also influences chemoresistance in AML patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73513

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Liu et al.
PloS one, 9(6), e98853-e98853 (2014-06-06)
Acute myeloid leukemia (AML), caused by abnormal proliferation and accumulation of hematopoietic progenitor cells, is one of the most common malignancies in adults. We reported here DYRK1A expression level was reduced in the bone marrow of adult AML patients, comparing
Chiara Di Vona et al.
Molecular cell, 57(3), 506-520 (2015-01-27)
DYRK1A is a dosage-sensitive protein kinase that fulfills key roles during development and in tissue homeostasis, and its dysregulation results in human pathologies. DYRK1A is present in both the nucleus and cytoplasm of mammalian cells, although its nuclear function remains
Karol Dowjat et al.
Journal of neuropathology and experimental neurology, 71(12), 1100-1112 (2012-11-14)
The triplication of the DYRK1A gene encoding proline-directed serine/threonine kinase and located in the critical region of Down syndrome (DS) has been implicated in cognitive deficits and intellectual disability of individuals with DS. We investigated the effect of abnormal levels
Elisa Giorgio et al.
Human mutation, 42(1), 102-116 (2020-12-01)
In genetic diseases, the most prevalent mechanism of pathogenicity is an altered expression of dosage-sensitive genes. Drugs that restore physiological levels of these genes should be effective in treating the associated conditions. We developed a screening strategy, based on a
N Janel et al.
Translational psychiatry, 4, e425-e425 (2014-08-15)
To determine whether apparent involvement of DYRK1A in Alzheimer's disease (AD) pathology makes it a candidate plasma biomarker for diagnosis, we developed a method to quantify plasma DYRK1A by immunoblot in transgenic mouse models having different gene dosages of Dyrk1a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service