Skip to Content
Merck
All Photos(2)

Key Documents

CWA-1080

Sigma-Aldrich

Anti-Human CD178/FasL (NOK-1) ColorWheel® Dye-Ready mAb

greener alternative

for use with ColorWheel® Dyes (Required, (Sold Separately))

Synonym(s):

Tumor necrosis factor ligand superfamily member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product CWA-1080 is not currently sold in your country. Contact Technical Service

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NOK-1, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 31.49 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG1κ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... FASLG(356)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone NOK-1 is a mouse monoclonal antibody that detects CD178 (FasL).

Immunogen

L5178Y mouse T lymphoma cells expressing recombinant human CD178.

Application

Quality Control Testing

Evaluated by Flow Cytometry in HEK293 cells transfected with CD178.

Staining of one million transiently CD178-transfected HEK 293 cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1080, Anti-Human CD178/FasL (NOK-1) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or a 1:1 mixture of an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-ready Allophycocyanin (APC) Dye.

Evaluated by Flow Cytometry in NK cells.

Flow Cytometry Analysis: Staining of one million Human NK cells was performed using 5 μL of a 1:1 mixture of Cat. No. CWA-1080, Anti-Human CD178 (NOK-1) ColorWheel® Dye-Ready mAb and Cat. No. CWD-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or an equivalent amount of Cat. No. CWA-1013 Mouse IgG1 isotype control and Cat. No. CWD-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

Tumor necrosis factor ligand superfamily member 6 (UniProt: P48023; also known as Apoptosis antigen ligand, APTL, CD95 ligand, CD95-L, Fas antigen ligand, Fas ligand, FasL, CD178) is encoded by the FASLG (also known as APT1LG1, CD95L, FASL, TNFSF6) gene (Gene ID: 356) in human. Fas Ligand is a single-pass, homotrimeric, type II membrane protein with a cytoplasmic domain (aa 1-80), a transmembrane domain (aa 81-102), and an extracellular domain (aa 103-281). The membrane-bound FasL (mFasL) undergoes two successive intramembrane proteolytic cleavages. The first one is processed by ADAM10, which produces an N-terminal fragment that lacks the receptor-binding extracellular domain. This ADAM10-processed FasL remnant form is still membrane anchored and further processed by signal peptide peptidase like 2A (SPPL2A) that liberates the FasL intracellular domain (FasL ICD). mFasL can also undergo proteolytic cleavage to produce a truncated soluble form (sFasL; aa 130-281). However, sFasL does not activate Fas, and it competes with the membrane bound FasL to reduce its cytotoxic activity. sFasL can bind to Fas, but does not induce its oligomerization, thereby preventing activation of the proapoptotic signaling pathway by the membrane bound FasL. FasL can also undergo N-glycosylation that enhances its apoptotic activity. Two isoforms of FasL have been described that are produced by alternative splicing. (Ref.: Volpe, E., et al. (2016). Front. Immunol. 7; 382; Schneider, P., et al. (1997). J. Biol. Chem. 272(30); 18827-18833).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 μL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 μL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

required but not provided

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service