Skip to Content
Merck
All Photos(1)

Key Documents

70088-M

Sigma-Aldrich

pBAC-3 DNA - Novagen

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$617.00

$617.00


Estimated to ship on28 May 2025



Select a Size

Change View
10 μG
$617.00

About This Item

UNSPSC Code:
41106612
NACRES:
NA.51

$617.00


Estimated to ship on28 May 2025


form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

storage temp.

−20°C

General description

pBAC-3 is a baculovirus transfer plasmid compatible with BacVector-1000, -2000, or -3000 Triple Cut Virus DNA for low background transfection and efficient utilization of the polh promoter and expression of secreted proteins. A 20 amino acid gp64 signal peptide sequence is capable of directing high levels of protein into the secretory pathway of infected insect cells. Cloning sites are provided for the creation of N-terminal fusions of an insert with His•Tag and/or S•Tag sequences. pBAC-3 provides an ATG start codon at the optimal position relative to native polyhedrin translation signals. The f1 origin is oriented so that infection with helper phage will produce virions containing single stranded DNA that corresponds to the coding strand. Single stranded sequencing of phage-derived DNA can be performed using the 1629DWN primer.

Warning

Toxicity: Standard Handling (A)

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service