CRISPR12H
CRISPR-Lenti Non-Target Control High Titer Transduction Particles
Select a Size
Select a Size
About This Item
Recommended Products
form
liquid
Quality Level
packaging
vial of 200 μL
concentration
5x108 VP/ml (via p24 assay)
application(s)
CRISPR
shipped in
dry ice
storage temp.
−70°C
Related Categories
Application
- Negative Control, CRISPR high titer lentivirus format.
To see more application data, protocols, vector maps visit sigma.com/crispr.
Features and Benefits
Other Notes
Physical form
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service